Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Obtusifolin-2-O-glucoside | 120163-18-0 | 446.40 | 5 MG
SDP

Catalog No. 5000416583
Encompass_Preferred

Obtusifolin-2-O-glucoside is a Tyrosinase inhibitor (IC50=9.2 μM) isolated from cassia seed. It inhibits melanin production in α-MSH-treated B16F10 melanoma cells at 10 μM for 48 hours. This product is for research use only.

  • Tyrosinase inhibitor (IC50=9.2 μM)
  • Isolated from cassia seed
  • Inhibits melanin production in melanoma cells
  • Purity: 95.30%
  • Appearance: Solid, yellow to brown
  • Molecular formula: C22H22O10
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (25 mg/mL)

Catalog No. 50-004-16583 Supplier Medchemexpress LLC Supplier No. HYN110635MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Obtusifolin-2-O-glucoside is a Tyrosinase inhibitor (IC50=9.2 μM) isolated from cassia seed. It inhibits melanin production in α-MSH-treated B16F10 melanoma cells at 10 μM for 48 hours. This product is for research use only.

  • Tyrosinase inhibitor (IC50=9.2 μM)
  • Isolated from cassia seed
  • Inhibits melanin production in melanoma cells
  • Purity: 95.30%
  • Appearance: Solid, yellow to brown
  • Molecular formula: C22H22O10
  • Shipping: Room temperature in continental US
  • Storage (powder): -20°C for 3 years
  • Storage (in solvent): -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (25 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.