Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ProSci OCC-1 Peptide
SDP

Catalog No. 16596619
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-596-619 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-596-619 Supplier ProSci Supplier No. 4997P0050
Only null left
Add to Cart
Add to Cart

Used for Blocking Activity

OCC-1 peptide is used for blocking the activity of OCC-1 antibody.

Specifications

Gene ID (Entrez) 387882
Species Human
Accession Number AAH61920
Common Name chromosome 12 open reading frame 75
Concentration 200 μg/mL
Content And Storage Store OCC-1 peptide (at -20°C, stable for one year.)
Format Liquid
Formulation PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide
For Use With (Application) Blocking Assay
Gene Symbol C12orf75
Quantity 0.05 mg
Sequence 16 amino acids near the amino terminus of human OCC-1
Synonym AGD3, OCC1, OCC-1, C12orf75, Overexpressed in colon carcinoma 1 protein, AGD3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.