Learn More
MSC OccuNomix Breathable Rain Bib Pants, Polyester, Yellow, Large, 1 ea
Catalog No. 5000766708
Encompass
These premium breathable rain bib pants are engineered for safety and performance, meeting ANSI/ISEA 107-2020 Class E standards and achieving Class 3 when paired with an ANSI shirt, vest, or jacket. Constructed from heavy-duty 300 Denier Polyester Oxford, they are water repellent and feature a breathable laminated membrane with sealed seams. Viral protection is also included, as they are ASTM F1671 Compliant.
  • Reflective tape, 2 in
  • Full-length 2-way waterproof zippers
  • Detachable black suspenders
  • Articulated knee design
  • Hook and loop adjustable ankle tabs
  • Hammer loop
  • Waterproof zip fly front with elastic waist
  • Three waterproof-zippered pockets (2 front, 1 back)
  • High-visibility
  • Waterproof construction
  • Adjustable waist and leg zippers
Catalog No. 50-007-66708 Supplier MSC Supplier No. 72946866
These premium breathable rain bib pants are engineered for safety and performance, meeting ANSI/ISEA 107-2020 Class E standards and achieving Class 3 when paired with an ANSI shirt, vest, or jacket. Constructed from heavy-duty 300 Denier Polyester Oxford, they are water repellent and feature a breathable laminated membrane with sealed seams. Viral protection is also included, as they are ASTM F1671 Compliant.
  • Reflective tape, 2 in
  • Full-length 2-way waterproof zippers
  • Detachable black suspenders
  • Articulated knee design
  • Hook and loop adjustable ankle tabs
  • Hammer loop
  • Waterproof zip fly front with elastic waist
  • Three waterproof-zippered pockets (2 front, 1 back)
  • High-visibility
  • Waterproof construction
  • Adjustable waist and leg zippers