Learn More
OccuNomix™ Cap Liners
Catalog No. 19520380 Shop All Occunomix International Inc Products
Click to view available options
Product Type:
Long-neck Basic Liner
Shoulder-Length Liner (Heavy-duty)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
19-520-380 Shoulder-Length Liner (Heavy-duty)
19-816-253 Long-neck Basic Liner
19-057-356 -
19-520-381 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-520-380 Supplier OccuNomix™ Supplier No. SS550
Add to Cart
Add to Cart
This item is not returnable. View return policy
Multiple styles from which to choose
  • Knitted Tube Liner: 100% polyester knit fits snugly over hardhat; folds up on outside of hat when not needed. Blocks wind tunneling inside crown of hat.
  • Heavy-Duty Shoulder-Length Liner: Shoulder-length cotton twill shell with sherpa lining. Two layers, warming pack pockets, elastic for snug fit, and insulated ear barrier.
  • Value Sherpa Liner: Shoulder length cotton twill shell with Sherpa lining.
  • Mid Length Liner: Single washed FR treated cotton outer shell with poly fleece lining. Two layers, warming pack pockets, reflective strip, elastic for snug fit and insulated ear barrier.
  • Shoulder Length Two-Way Liner: 100% cotton top with polyester-fleece lining; removable 100% polyester bottom. Fleece and polyester are flame resistant.
  • Extra-Long Liner with Removable Mouthpiece: Two layers with navy shell and red lining with extra-long neck length.
  • Three-Layer Shoulder-Length Liner: 100% cotton twill shell with Thinsulate™ thermal middle layer and heavyweight polyester-fleece lining.
  • Specifications
    Product Type Shoulder-Length Liner (Heavy-duty)