Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OccuNomix™ Embossed Rubber Back Anti-Vibration Gloves with VEP Padding

Catalog No. 19815559 Shop All Occunomix International Inc Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-815-559 2X-Large
19-815-555 Small
19-815-556 Medium
19-815-557 Large
19-815-558 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-815-559 Supplier OccuNomix™ Supplier No. 426066
This item is not returnable. View return policy

Gloves feature embossed rubber on the top of the hand for knuckle protection, while a spandex back provides a secure fit.

  • Premium leather palm
  • 4-way stretch spandex back
  • Visco elastic polymer padding
  • Hook & Loop wrist closure
  • Terry cloth thumb to wipe away sweat

Specifications

Back Material Spandex
Color Black
Hand Specific
Material Synthetic Leather
Product Type Anti-vibration Gloves
Size 2X-Large
For Use With (Application) Medium to Heavy Industrial Power Tools
Glove Cuff Style Hook and Loop
Series 426
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.