Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OccuNomix™ Half-Finger Terry Cloth Anti-Vibration Gloves

Catalog No. 19816308 Shop All Occunomix International Inc Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-816-308 2X-Large
19-816-304 Small
19-816-305 Medium
19-816-306 Large
19-816-307 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-816-308 Supplier OccuNomix™ Supplier No. 422066
This item is not returnable. View return policy

Terry Cloth back material absorbs perspiration, while VEP gel padding provides protection from impact and vibration.

  • VEP gel padding covers the palm, index finger, and lower thumb
  • Optimally suited for impact power tools, sanders, and small power tools
  • Hook and loop closure

Specifications

Color Blue, Gray
Hand Specific
Material Terrycloth
Product Type Anti-vibration Gloves
Size 2X-Large
For Use With (Application) Power Tools and Sanders
Glove Cuff Style Hook and Loop
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.