Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OccuNomix™ Deluxe Knee Pad

Catalog No. 19057351 Shop All Occunomix International Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Closure Type:
Hook and loop
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
19-057-351 Hook and loop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-057-351 Supplier OccuNomix™ Supplier No. 127
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Black or white
Closure Type Hook and loop
Product Type Classic™ Knee Pad
For Use With (Application) Delicate surfaces
Includes Non-marring molded Rubber cap with gripping ridges, adjustable straps
Liner Material EVA Foam
Material Polyester and tricot Fabrics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.