Learn More
MSC OccuNomix Flame Resistant Gaiter, Modacrylic Blend, Universal, Navy, 1 ea
Catalog No. 5000673738
Encompass
This flame resistant modacrylic multi-functional gaiter provides protection and comfort. Designed to be lightweight with maximum stretch, it can be worn alone or under headwear for versatility.
  • Lightweight and maximum stretch for comfortable fit
  • Can be worn alone or under hard hat or helmet
  • Meets ASTM F1506
  • Arc rating: ATPV = 5.8 cal/cm2
  • NFPA 70E/CAT 1 compliant
  • Constructed from 7.5 oz. modacrylic/cotton/spandex jersey knit with Nomex® thread
Catalog No. 50-006-73738 Supplier MSC Supplier No. 46028106
This flame resistant modacrylic multi-functional gaiter provides protection and comfort. Designed to be lightweight with maximum stretch, it can be worn alone or under headwear for versatility.
  • Lightweight and maximum stretch for comfortable fit
  • Can be worn alone or under hard hat or helmet
  • Meets ASTM F1506
  • Arc rating: ATPV = 5.8 cal/cm2
  • NFPA 70E/CAT 1 compliant
  • Constructed from 7.5 oz. modacrylic/cotton/spandex jersey knit with Nomex® thread