Learn More
MSC OccuNomix Hi-Vis Cold Weather Jacket, Large, Polyester, 1 ea
Catalog No. 5000792791
Encompass
This jacket is designed for cold weather and high visibility applications. It is constructed from polyester, featuring a zipper closure and a convenient roll-away hood. The jacket is equipped with six pockets and provides waterproof protection, along with two mic tabs and a removable lining. It meets ANSI/ISEA 107-2015 Type R, Class 3 standards for safety.
  • Two in. silver reflective tape
  • One hundred percent ANSI polyester with polyurethane (PU) coating and sealed seams for waterproofing
  • One hundred percent black polyester fleece zip-out liner that can be worn as a separate jacket
  • Black bottom front, collar, cuffs, and waistband
  • Roll-away hood
  • Two mic tabs
  • Removable lining
  • Six pockets
Catalog No. 50-007-92791 Supplier MSC Supplier No. 40520256
This jacket is designed for cold weather and high visibility applications. It is constructed from polyester, featuring a zipper closure and a convenient roll-away hood. The jacket is equipped with six pockets and provides waterproof protection, along with two mic tabs and a removable lining. It meets ANSI/ISEA 107-2015 Type R, Class 3 standards for safety.
  • Two in. silver reflective tape
  • One hundred percent ANSI polyester with polyurethane (PU) coating and sealed seams for waterproofing
  • One hundred percent black polyester fleece zip-out liner that can be worn as a separate jacket
  • Black bottom front, collar, cuffs, and waistband
  • Roll-away hood
  • Two mic tabs
  • Removable lining
  • Six pockets