Learn More
MSC OccuNomix Hi-Vis Vest, 4X/5X, Yellow, ANSI 2, 1 ea
Catalog No. 5000766355
Encompass
This high visibility vest is designed for safety in various work environments. Featuring a 5-point breakaway design, it provides enhanced protection and meets stringent safety standards for professional use.
  • Hi-visibility yellow color for increased visibility
  • Hook & loop closure for easy wear and removal
  • Two pockets for convenient storage
  • Chest size of 66 in.
  • Constructed from mesh material for breathability
  • Equipped with one 2 in. wide silver horizontal stripe
  • Meets ANSI Class 2, ISEA 107 Class 2, Type R, and ANSI 107-2010 approval listings
  • Available in 4X/5X-Large sizes
Catalog No. 50-007-66355 Supplier MSC Supplier No. 96552823
This high visibility vest is designed for safety in various work environments. Featuring a 5-point breakaway design, it provides enhanced protection and meets stringent safety standards for professional use.
  • Hi-visibility yellow color for increased visibility
  • Hook & loop closure for easy wear and removal
  • Two pockets for convenient storage
  • Chest size of 66 in.
  • Constructed from mesh material for breathability
  • Equipped with one 2 in. wide silver horizontal stripe
  • Meets ANSI Class 2, ISEA 107 Class 2, Type R, and ANSI 107-2010 approval listings
  • Available in 4X/5X-Large sizes