Learn More
MSC OccuNomix Hi-Vis Vest, ANSI Class 2, 3X, 1 ea.
Catalog No. 5000767383
Encompass
The OccuNomix Classic Two-Tone Surveyor's Vest features lightweight 100% polyester construction with a mesh back and tricot front. It includes 2 in silver reflective tape, with one 360° horizontal and two vertical stripes for visibility. Equipped with 10 pockets (2 inside lower, 2 outside lower flapped, 4 pen, 1 divided), it also has large arm openings for outerwear and a zipper closure. This vest meets ANSI 107 Type R Class 2 standards.
  • Constructed from 100% ANSI polyester tricot front and mesh back
  • Silver reflective tape with one 360° horizontal stripe and two vertical stripes backed by ANSI compliant trim
  • Large arm opening for use over cold weather outerwear
  • Equipped with 10 pockets, including 2 inner lower, 2 outer lower flapped, 4 pen, and 1 divided into 2 sections
  • Zipper closure
Catalog No. 50-007-67383 Supplier MSC Supplier No. 54202783
The OccuNomix Classic Two-Tone Surveyor's Vest features lightweight 100% polyester construction with a mesh back and tricot front. It includes 2 in silver reflective tape, with one 360° horizontal and two vertical stripes for visibility. Equipped with 10 pockets (2 inside lower, 2 outside lower flapped, 4 pen, 1 divided), it also has large arm openings for outerwear and a zipper closure. This vest meets ANSI 107 Type R Class 2 standards.
  • Constructed from 100% ANSI polyester tricot front and mesh back
  • Silver reflective tape with one 360° horizontal stripe and two vertical stripes backed by ANSI compliant trim
  • Large arm opening for use over cold weather outerwear
  • Equipped with 10 pockets, including 2 inner lower, 2 outer lower flapped, 4 pen, and 1 divided into 2 sections
  • Zipper closure