Learn More
MSC OccuNomix High Visibility Vest, 2X-Large, Yellow, 1 ea
Catalog No. 5000794233
Encompass
This high visibility vest is designed for safety in various work environments. Made from mesh material, it features a hook and loop closure for easy wear and two pockets for convenience.
  • Size: 2X-Large
  • Color: Yellow
  • Closure type: Hook & loop
  • Pockets: 2
  • Material: Mesh
  • Horizontal stripes: 1, silver, 2 in wide
  • ANSI Class 2 approved
  • Complies with ANSI 107-2015 standards
Catalog No. 50-007-94233 Supplier MSC Supplier No. 39098421
This high visibility vest is designed for safety in various work environments. Made from mesh material, it features a hook and loop closure for easy wear and two pockets for convenience.
  • Size: 2X-Large
  • Color: Yellow
  • Closure type: Hook & loop
  • Pockets: 2
  • Material: Mesh
  • Horizontal stripes: 1, silver, 2 in wide
  • ANSI Class 2 approved
  • Complies with ANSI 107-2015 standards