Learn More
MSC OccuNomix High Visibility Vest, X-Large, Yellow, Class 2, 1 ea
Catalog No. 5000794007
Encompass
This high visibility vest provides essential safety for workers in environments requiring enhanced visibility. Designed for demanding use, it features a durable mesh construction, multiple pockets for utility, and adheres to stringent safety standards.
  • X-Large size
  • Hi-visibility yellow color
  • Zipper closure
  • Seven pockets
  • Mesh material
  • Two silver horizontal stripes, 2 in wide
  • Fall protection D-ring
  • Mic tab
  • Withstands 25 washes
  • ANSI Class 2 approved
  • Meets ISEA 107-2015 Type R Class 2 and ANSI/ISEA 107-2015 Type R, Class 2 standards
Catalog No. 50-007-94007 Supplier MSC Supplier No. 96552674
This high visibility vest provides essential safety for workers in environments requiring enhanced visibility. Designed for demanding use, it features a durable mesh construction, multiple pockets for utility, and adheres to stringent safety standards.
  • X-Large size
  • Hi-visibility yellow color
  • Zipper closure
  • Seven pockets
  • Mesh material
  • Two silver horizontal stripes, 2 in wide
  • Fall protection D-ring
  • Mic tab
  • Withstands 25 washes
  • ANSI Class 2 approved
  • Meets ISEA 107-2015 Type R Class 2 and ANSI/ISEA 107-2015 Type R, Class 2 standards