Learn More
OccuNomix™ Hot Rods Warming Pack
Catalog No. 19057299 Shop All Occunomix International Inc Products
Click to view available options
Color:
Orange
Catalog No. Color
19-057-299 Orange
1 options
Catalog No. 19-057-299 Supplier OccuNomix™ Supplier No. 110010R
Includes
Pack of 10 (5 pairs)
Warming packs offer all-day comfort in cold temperature applications
Packs warm up within two minutes and feature a new and improved formula.
  • Small enough to slip into your gloves
  • Last up to 8 hours with an average temperature of 120°F (48.9°C)
Specifications
Color Orange
Disposable Single-use
Product Type Hand Warming Packs
Quantity Pack of 10 (5 pairs)