Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OccuNomix™ Hot Rods Winter Liners: Shoulder Length

Catalog No. 19816256 Shop All Occunomix International Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Material:
Cotton
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-816-256 Cotton
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-816-256 Supplier OccuNomix™ Supplier No. SB505
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Feature two built-in pockets to hold regular-size Hot Rods™ warming packs

  • Short elastic strip in the forehead area keeps liner snug to face
  • Insulated ear barriers provide extra protection against wind, water and cold
  • Strip of reflective film above the back of neck offers visibility in low-light or dark hours of the day

  • Models RB405 and SB405
    • Two layers
    • 100% cotton twill top
    • Fleece lining
    • Flame-retardant until washed
    • Color: Navy
    Model RQ300
    • Three layers
    • 100% quilted nylon shell
    • 2mm foam center layer
    • Fleece lining
    • Color: Green

    Specifications

    Material Cotton
    Color Navy
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.