Learn More
MSC OccuNomix Mesh Vest, ANSI Class 2, Yellow, 4X/5X, 1 ea
Catalog No. 5000794326
Encompass
This Mesh Standard Vest is designed for enhanced worker visibility and safety. Constructed from durable polyester mesh, it meets ANSI/ISEA 107-2020 Type R Class 2 standards for high-visibility apparel.
  • Two-inch reflective tape
  • Hook and loop closure
  • One pocket
  • One inner left chest pocket
  • Available in sizes S/M through 4X/5X
Catalog No. 50-007-94326 Supplier MSC Supplier No. 56185978
This Mesh Standard Vest is designed for enhanced worker visibility and safety. Constructed from durable polyester mesh, it meets ANSI/ISEA 107-2020 Type R Class 2 standards for high-visibility apparel.
  • Two-inch reflective tape
  • Hook and loop closure
  • One pocket
  • One inner left chest pocket
  • Available in sizes S/M through 4X/5X