Learn More
OccuNomix™ MiraCool™ Bandanas
Catalog No. 19057352 Shop All Occunomix International Inc Products
Click to view available options
Color:
Assorted
Jungle
Khaki
Red
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-057-352 Red
19-130-2381 Assorted
19-057-353 Khaki
19-520-309 Jungle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-057-352 Supplier OccuNomix™ Supplier No. 940038
Add to Cart
Add to Cart
Lightweight neck bandanas with cooling power and the comfort of cotton
  • 100 percent cotton with cooling crystals
  • Works without freezing or ice
  • Simply soak in cold water for 10 to 20 minutes
  • Stay hydrated for days
  • Reusable
  • Triple washed to prevent fabric bleeding
  • Flame-resistant, arc-rated, treated cotton (treatment lasts for up to 50 washes)
Specifications
Material Cotton
Product Type Cooling Bandana
Size Universal
Color Red
Closure Type Tie
Includes Cooling Crystals