Learn More
Description
- 100% cotton
- Super-absorbent cooling crystals work without ice or freezing
- Simply soak in cold water – stays hydrated for days
- Hook-and-loop front closures and expandable sides
- One size fits all
- Color: High-visibility orange
Specifications
Specifications
| Material | Cotton |
| Product Type | Cooling Vest |
| Size | Regular |
| Color | Hi-vis Orange |
| For Use With (Application) | For construction site safety equipment comfort |
| Quantity | Each |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.