Learn More
MSC OccuNomix Rain Pants, Polyester Oxford, Hi-Vis Yellow, X-Large, 1 ea
Catalog No. 5000794026
Encompass
These rain pants are designed for high-visibility applications, offering breathability and waterproof protection with sealed seams for comfort and safety in wet conditions.
  • Constructed from 150 Denier Polyester Oxford with PU coating
  • Waterproof with sealed seams
  • Meets ANSI/ISEA 107-2020 Class E standards
  • Features 2 in. reflective tape
  • Adjustable snap leg opening
  • Includes 3 pockets: 2 pass-through and 1 right side leg expandable with hook and loop flap
Catalog No. 50-007-94026 Supplier MSC Supplier No. 54645759
These rain pants are designed for high-visibility applications, offering breathability and waterproof protection with sealed seams for comfort and safety in wet conditions.
  • Constructed from 150 Denier Polyester Oxford with PU coating
  • Waterproof with sealed seams
  • Meets ANSI/ISEA 107-2020 Class E standards
  • Features 2 in. reflective tape
  • Adjustable snap leg opening
  • Includes 3 pockets: 2 pass-through and 1 right side leg expandable with hook and loop flap