Learn More
MSC Occunomix Rain Pants, Polyester, Reflective, 2X-Large, 1 ea
Catalog No. 5000782007
Encompass
Occunomix premium breathable rain pants provide protection and visibility. Made from 100% ANSI 107-2010 polyester 300 denier with a waterproof membrane and sealed seams, they feature 2-inch reflective material. Designed for comfort with full-length zippers, a CORDURA back patch, and gusseted knees, they are Class E compliant.
  • Waterproof construction with sealed seams.
  • Full-length 2-way waterproof zippers for waist to ankle access.
  • CORDURA back patch provides stability on wet equipment.
  • Gusseted knee design provides comfort and mobility.
  • Two front pockets and one back pocket with waterproof zippers.
  • Includes 2-inch reflective material with four 360° horizontal leg stripes.
  • Constructed from 100% ANSI 107-2010 polyester 300 denier breathable waterproof membrane.
  • Class E compliant.
Catalog No. 50-007-82007 Supplier MSC Supplier No. 72946700
Occunomix premium breathable rain pants provide protection and visibility. Made from 100% ANSI 107-2010 polyester 300 denier with a waterproof membrane and sealed seams, they feature 2-inch reflective material. Designed for comfort with full-length zippers, a CORDURA back patch, and gusseted knees, they are Class E compliant.
  • Waterproof construction with sealed seams.
  • Full-length 2-way waterproof zippers for waist to ankle access.
  • CORDURA back patch provides stability on wet equipment.
  • Gusseted knee design provides comfort and mobility.
  • Two front pockets and one back pocket with waterproof zippers.
  • Includes 2-inch reflective material with four 360° horizontal leg stripes.
  • Constructed from 100% ANSI 107-2010 polyester 300 denier breathable waterproof membrane.
  • Class E compliant.