Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC OccuNomix Seat Belt Cover, Yellow, Polyester, 18.5 in., 1 ea

Catalog No. 5000766045
Encompass

This polyester seat belt cover is designed for comfort and visibility. It is suitable for use with almost any car seat belt.

  • Type: Seat belt cover
  • For use with: Almost every car
  • Color: Yellow
  • Material: Polyester
  • Length: 18½ in.
  • Number of pockets: 0

Catalog No. 50-007-66045 Supplier MSC Supplier No. 89522429
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This polyester seat belt cover is designed for comfort and visibility. It is suitable for use with almost any car seat belt.

  • Type: Seat belt cover
  • For use with: Almost every car
  • Color: Yellow
  • Material: Polyester
  • Length: 18½ in.
  • Number of pockets: 0

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.