Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OccuNomix™ Terry Toppers™ Snap-On Sweatbands for Hard Hats

Catalog No. 19057354 Shop All Occunomix International Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Color:
Beige
Navy Blue
Material:
Terrycloth
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material
19-057-354 Navy Blue -
19-057-401 Beige Terrycloth
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-057-354 Supplier OccuNomix™ Supplier No. 870B10001
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

100% Super-sorbing soft terry

  • Absorbs without needing to be washed first
  • Easy snap-on application
  • Helps headgear fit more securely
  • No foam middle
  • Machine washable

Specifications

Color Navy Blue
For Use With (Equipment) Hard Hats
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.