Learn More
MSC OccuNomix Vest, 5-Point Breakaway, ANSI Class 2, X-Large, 1 ea
Catalog No. 5000794071
Encompass
The OccuNomix ECO-IMB2T is a high-visibility, two-tone mesh safety vest designed for workers operating in roadway, construction, and industrial environments where visibility and snag-away safety features are required. It enhances worker safety by providing increased visibility and a break-away system.
  • Meets ANSI/ISEA 107-2020 Class 2 - Type R
  • Polyester mesh construction
  • Two-inch reflective tape with contrasting trim
  • Quick release zipper closure
  • Five-point break-away design
  • Two pockets: 1 left chest, 1 inner lower
Catalog No. 50-007-94071 Supplier MSC Supplier No. 39790647
The OccuNomix ECO-IMB2T is a high-visibility, two-tone mesh safety vest designed for workers operating in roadway, construction, and industrial environments where visibility and snag-away safety features are required. It enhances worker safety by providing increased visibility and a break-away system.
  • Meets ANSI/ISEA 107-2020 Class 2 - Type R
  • Polyester mesh construction
  • Two-inch reflective tape with contrasting trim
  • Quick release zipper closure
  • Five-point break-away design
  • Two pockets: 1 left chest, 1 inner lower