Learn More
MSC OccuNomix Vest, Hi-Vis, Large Yellow, ANSI Class 2, 1 ea
Catalog No. 5000795086
Encompass
This high visibility vest features a hook & loop closure and two pockets. Designed for enhanced safety, it provides high visibility and meets ANSI 107-2015 standards for workplace safety.
  • Mesh material composition
  • Large size, chest 40–42 in
  • Hook & loop closure
  • Two pockets
  • One horizontal silver reflective stripe, 2 in wide
  • ANSI Class 2 approved
Catalog No. 50-007-95086 Supplier MSC Supplier No. 39098389
This high visibility vest features a hook & loop closure and two pockets. Designed for enhanced safety, it provides high visibility and meets ANSI 107-2015 standards for workplace safety.
  • Mesh material composition
  • Large size, chest 40–42 in
  • Hook & loop closure
  • Two pockets
  • One horizontal silver reflective stripe, 2 in wide
  • ANSI Class 2 approved