Learn More
MSC OccuNomix Winter Liner, Navy Blue, Flame Resistant, 1 ea
Catalog No. 5000766056
Encompass
This Navy Blue, Mid Length Winter Liner features a 2-layer construction and is made of cotton. It is designed to be flame resistant, meeting NFPA 70E and ASTM F1506 standards, making it suitable for demanding industrial applications.
  • Provides effective cold weather protection as a winter liner
  • Features a mid-neck length for extended coverage
  • Constructed from durable cotton material
  • Flame resistant for enhanced safety
  • Equipped with a hook & loop adjustment for a secure fit
  • Designed for universal sizing to fit most users
  • Compatible for use over a brim
  • Meets NFPA 70E and ASTM F1506 safety standards
  • Two-layer construction for added warmth
Catalog No. 50-007-66056 Supplier MSC Supplier No. 91395798
This Navy Blue, Mid Length Winter Liner features a 2-layer construction and is made of cotton. It is designed to be flame resistant, meeting NFPA 70E and ASTM F1506 standards, making it suitable for demanding industrial applications.
  • Provides effective cold weather protection as a winter liner
  • Features a mid-neck length for extended coverage
  • Constructed from durable cotton material
  • Flame resistant for enhanced safety
  • Equipped with a hook & loop adjustment for a secure fit
  • Designed for universal sizing to fit most users
  • Compatible for use over a brim
  • Meets NFPA 70E and ASTM F1506 safety standards
  • Two-layer construction for added warmth