Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Ocean Pacific™ Lab Master™ Nitrile Medical Examination Gloves with Textured Fingertips

Catalog No. 296366820 Shop All Medicom Products
Change view
Click to view available options
Size:
X-Small
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
29-636-6820 X-Small
29-636-6822 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-636-6820 Supplier Medicom™ Supplier No. OPXSLM2
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ocean Pacific™ Lab Master™ Nitrile Medical Examination Gloves with Textured Fingertips. Provides the protection and strength of nitrile without the chemical accelerators commonly found in nitrile gloves. Textured fingertips for a secure grip, even when wet. Chemo-tested.

  • These versatile all-purpose gloves provide all the protection and strength of nitrile without the chemical accelerators commonly found in nitrile gloves.
  • The DermaSense technology eliminates accelerators to reduce the risk of skin reactions.
  • Chemo-tested, accelerator-free, safe for food handling.
  • Designed for wider palm, shorter fingers.

Specifications

Chemotherapeutic Rated Yes
Chlorinated No
Accelerator Free Yes
Applications Healthcare, Industrial, Laboratory
AQL 1.5
Color Indigo
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 2.76 mil
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small
Sterility Non-sterile
Thickness (English) 2.76 mil
Thickness (Metric) 0.07 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Quantity 200/Box
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.