Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ Ocean Pacific™ Ninja™ Nitrile Medical Examination Gloves

Catalog No. 19999742 Shop All Medicom Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-999-742 Small
19-999-743 Medium
19-999-744 Large
19-999-745 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-999-742 Supplier Medicom™ Supplier No. OPSNI
Add to Cart
Add to Cart
This item is not returnable. View return policy

Nitrile Medical Examination Gloves are thicker, fully textured and provide optimal grip, puncture resistance and protection against bloodborne pathogens, chemical lubricants and sharp instruments. Chemo tested and fentanyl safe.

  • Fully textured glove provides optimal grip, puncture resistance and protection against bloodborne pathogens, chemical lubricants and sharp instruments.
  • Chemo tested and fentanyl safe.
  • Comfortable fit. Nitrile gloves conform to hands, providing a snug, comfortable fit to reduce hand fatigue.
  • Optimal stretch. Nitrile gloves offer optimal flexibility, making them ideal for procedures that require manual dexterity and tactile sensitivity.
  • Latex free. Nitrile gloves are not made with natural rubber latex, so they are safe for professionals and patients with a latex allergy or sensitivity.

Specifications

Chlorinated Yes
Accelerator Free No
Applications Laboratory, Healthcare, Industrial
AQL 2.5
Color Black
Disposable Single-use
Exterior Finish Fully Textured
Finger Thickness (English) 4.72 mil.
Finger Thickness (Metric) 0.12 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.94 mil.
Palm Thickness (Metric) 0.1 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size Small
Sterility Non-sterile
Thickness (English) 3.94 mil.
Thickness (Metric) 0.1 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.