Learn More
Aviva Systems Biology Corp OCEL1 Antibody - C-terminal region (ARP50679_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50679_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OCEL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human OCEL1. Alternate names: FLJ22709, FWP009, S863-9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Pig, Rat
Catalog No. 50-014-60409