Learn More
AdipoGen OCH (250 µg)
SDP
Supplier:  AdipoGen AGCR13593C250
Encompass_Preferred
Chemical. CAS: 383187-82-4. Formula: C39H77NO9. MW: 704. Truncated analog of alpha-GalCer. iNKT cell glycolipid agonist. Immunosuppressive. Th2 polarizing. Stimulation of NKT with OCH results in release of primarily Th2 cytokine compared to stimulation with alpha-GalCer, which results in release of a mixture of Th1 and Th2 cytokines. Induces stronger interleukin-4 (IL-4) secretion and weaker interferon-gamma (IFN-gamma) secretion than alpha-GalCer both in vitro and in vivo. Inducer of interleukin-10 (IL-10) and suppressor of diabetes. Shown to prevent experimental autoimmune encephalomyelitis (EAE) in mice, an animal model of multiple sclerosis. Has beneficial effects on type I diabetes in NOD mice. Suppressor of collagen-induced arthritis. Might be useful for immunotherapy of transplant rejection.
Catalog No. 50-205-3959