Learn More
Cayman Chemical Ochratoxin A, Purity: ≥98%, CAS: 303-47-9, 10 mg
SDP
Catalog No. 502954932
Encompass_Preferred
A mycotoxin and an active metabolite of ochratoxin C; increases renal lipid peroxide levels and the number of apoptotic cells, as well as reduces renal SOD activity in rats at 120 μg/kg; topical application induces DNA damage, cell cycle arrest at the G0/G1 phase, and apoptosis in mouse skin cells; initiates tumor formation in a two-stage mouse skin tumorigenesis model; has been found in food products and poultry feed
Catalog No. 50-295-4932 Supplier Cayman Chemical Supplier No. 1143910 MG
A mycotoxin and an active metabolite of ochratoxin C; increases renal lipid peroxide levels and the number of apoptotic cells, as well as reduces renal SOD activity in rats at 120 μg/kg; topical application induces DNA damage, cell cycle arrest at the G0/G1 phase, and apoptosis in mouse skin cells; initiates tumor formation in a two-stage mouse skin tumorigenesis model; has been found in food products and poultry feed