Learn More
Cayman Chemical Ochratoxin A-13C20, Purity: ≥98%, CAS: 911392-42-2, 1.2 mL
SDP
Catalog No. 502991851
Encompass_Preferred
An internal standard for the quantification of ochratoxin A by GC- or LC-MS
Catalog No. 50-299-1851 Supplier Cayman Chemical Supplier No. 312951.2 ML
An internal standard for the quantification of ochratoxin A by GC- or LC-MS