Learn More
Enzo Life Sciences Ochratoxin A (5mg). CAS: 303-47-9
SDP
Catalog No. 502009617
Encompass_Preferred
Mycotoxin. Natural contaminant of mouldy food and feed. It has a number of toxic effects, the most prominent being nephrotoxicity. Immunosuppressive, genotoxic, teratogenic and carcinogenic. Stimulates lipid peroxidation. Induces oxidative DNA lesions coupled with direct DNA adducts via quinone formation. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in methanol (10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9617 Supplier Enzo Life Sciences Supplier No. ALX630089M005
Mycotoxin. Natural contaminant of mouldy food and feed. It has a number of toxic effects, the most prominent being nephrotoxicity. Immunosuppressive, genotoxic, teratogenic and carcinogenic. Stimulates lipid peroxidation. Induces oxidative DNA lesions coupled with direct DNA adducts via quinone formation. Purity: ≥98% (HPLC, TLC). Solubility: Soluble in methanol (10mg/ml). Long Term Storage: +4°C.