Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ochratoxin A Solution, 10 μg/mL in Acetonitrile, MilliporeSigma™ Supelco™

Catalog No. 111018151 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8151 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8151 Supplier MilliporeSigma Supelco Supplier No. 340372MLR
Add to Cart
Add to Cart

Certan Vial

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 303-47-9
Linear Formula C20H18 ClNO6
MDL Number MFCD00078079
Solubility Soluble in methanol
Type Analytical Standard
Quantity 2 mL
Formula Weight 403.81
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 10 μg/mL in acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.