Learn More
Cayman Chemical Ochratoxin B-13C20, Purity: ≥98%, 1.2 mL
SDP
Catalog No. 502942369
Encompass_Preferred
An internal standard for the quantification of ochratoxin B by GC- or LC-MS
Catalog No. 50-294-2369 Supplier Cayman Chemical Supplier No. 402381.2 ML
An internal standard for the quantification of ochratoxin B by GC- or LC-MS