Learn More
Aviva Systems Biology Corp OCIAD1 Antibody - C-terminal region (ARP48378_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48378_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OCIAD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human OCIAD1. Alternate names: Asrij, FLJ20455, MGC111072, OCIA, TPA018, ASRIJApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59807