Learn More
Aviva Systems Biology Corp OCIAD2 Antibody - middle region (ARP52941_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52941_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against OCIAD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human OCIAD2. Alternate names: DKFZp686C03164, MGC45416ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60874