Learn More
Aviva Systems Biology Corp Ociad2 Antibody - N-terminal region (ARP52940_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52940_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Ociad2. It was validated on Western Blot. . Alternate names: 1810027I20Rik, MGC51598ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-285-790