Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ocifisertib (fumarate) | 1616420-30-4 | 99.6% | 650.7 g/mol | C37H38N4O7 | 10MM 1ML
SDP

Catalog No. 5000238783
Encompass_Preferred

Ocifisertib (CFI-400945) fumarate is a potent, selective inhibitor of polo-like kinase 4 (PLK4) used in preclinical and in vitro research. It exhibits subnanomolar binding affinity (Ki ≈ 0.26 nM) and low-nanomolar cellular potency (PLK4 IC50 ≈ 2.8 nM), and has demonstrated antiproliferative activity in multiple cancer cell models.

  • Potent PLK4 inhibition with Ki ≈ 0.26 nM and PLK4 IC50 ≈ 2.8 nM.
  • Suitable for biochemical kinase assays and cell-based proliferation studies.
  • Provided as a 10 mM solution in 1 mL aliquots for convenient screening use.
  • Fumarate salt form to support handling and solubility.
  • Molecular formula C37H38N4O7; molecular weight 650.7 g/mol.
  • High reported purity from third-party suppliers; verify lot certificate before use.

Catalog No. 50-002-38783 Supplier Medchemexpress LLC Supplier No. HY12300B10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ocifisertib (CFI-400945) fumarate is a potent, selective inhibitor of polo-like kinase 4 (PLK4) used in preclinical and in vitro research. It exhibits subnanomolar binding affinity (Ki ≈ 0.26 nM) and low-nanomolar cellular potency (PLK4 IC50 ≈ 2.8 nM), and has demonstrated antiproliferative activity in multiple cancer cell models.

  • Potent PLK4 inhibition with Ki ≈ 0.26 nM and PLK4 IC50 ≈ 2.8 nM.
  • Suitable for biochemical kinase assays and cell-based proliferation studies.
  • Provided as a 10 mM solution in 1 mL aliquots for convenient screening use.
  • Fumarate salt form to support handling and solubility.
  • Molecular formula C37H38N4O7; molecular weight 650.7 g/mol.
  • High reported purity from third-party suppliers; verify lot certificate before use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.