Learn More
Medchemexpress LLC Ocifisertib fumarate | 1616420-30-4 | 99.3% | 650.72 Da | C37H38N4O7 | 5MG
SDP
Catalog No. 5000235725
Encompass_Preferred
Ocifisertib fumarate is the fumarate salt of Ocifisertib (CFI-400945), a potent, selective, orally bioavailable inhibitor of polo-like kinase 4 (PLK4) used as a research tool in oncology and cell biology to study centriole duplication and mitotic defects. Reported potency values support activity in biochemical and cellular assays.
Catalog No. 50-002-35725 Supplier Medchemexpress LLC Supplier No. HY12300B5MG
Ocifisertib fumarate is the fumarate salt of Ocifisertib (CFI-400945), a potent, selective, orally bioavailable inhibitor of polo-like kinase 4 (PLK4) used as a research tool in oncology and cell biology to study centriole duplication and mitotic defects. Reported potency values support activity in biochemical and cellular assays.