Learn More
Medchemexpress LLC Ocifisertib (fumarate) | 1616420-30-4 | 99.3% | 650.72 g/mol | C37H38N4O7 | 100MG
SDP
Catalog No. 5000238532
Encompass_Preferred
Ocifisertib (fumarate) is the fumarate salt form of a potent and selective ATP-competitive inhibitor of polo-like kinase 4 (PLK4) supplied for laboratory research. It is provided as a solid research reagent with molecular formula C37H38N4O7 and molecular weight 650.72 g/mol.
Catalog No. 50-002-38532 Supplier Medchemexpress LLC Supplier No. HY12300B100MG
Ocifisertib (fumarate) is the fumarate salt form of a potent and selective ATP-competitive inhibitor of polo-like kinase 4 (PLK4) supplied for laboratory research. It is provided as a solid research reagent with molecular formula C37H38N4O7 and molecular weight 650.72 g/mol.