Learn More
Medchemexpress LLC Ocifisertib (fumarate) | 1616420-30-4 | 99.3% | 650.72 g/mol | C37H38N4O7 | 10MG
SDP
Catalog No. 5000235521
Encompass_Preferred
Ocifisertib fumarate is the fumarate salt of CFI-400945, an orally bioavailable small-molecule inhibitor of Polo-like kinase 4 (PLK4) for preclinical research. It is supplied as a powder for in vitro and in vivo studies and shows low-nanomolar potency against PLK4 and activity against several related kinases.
Catalog No. 50-002-35521 Supplier Medchemexpress LLC Supplier No. HY12300B10MG
Ocifisertib fumarate is the fumarate salt of CFI-400945, an orally bioavailable small-molecule inhibitor of Polo-like kinase 4 (PLK4) for preclinical research. It is supplied as a powder for in vitro and in vivo studies and shows low-nanomolar potency against PLK4 and activity against several related kinases.