Learn More
Cayman Chemical OclacItInIb 10mg
SDP
Catalog No. 502979388
Encompass_Preferred
A JAK family kinase inhibitor (IC50s = 10, 18, 99, and 84 nM for JAK1, JAK2, JAK3, and TYK2, respectively); selective for JAK kinases over a panel of 38 additional kinases at 1 μM; inhibits LPS-induced increases in IL-12 and TNF-α levels in murine BMDCs in a concentration-dependent manner; topical administration reduces scratching behavior and EAr edema, as well as decreases levels of IL-1β, IL-4, and IL-6 in EAr skin, in a mouse model of TDI-induced allergic dermatitis at 0.1, 0.25, and 0.5%
Catalog No. 50-297-9388 Supplier Cayman Chemical Supplier No. 1872210 MG
A JAK family kinase inhibitor (IC50s = 10, 18, 99, and 84 nM for JAK1, JAK2, JAK3, and TYK2, respectively); selective for JAK kinases over a panel of 38 additional kinases at 1 μM; inhibits LPS-induced increases in IL-12 and TNF-α levels in murine BMDCs in a concentration-dependent manner; topical administration reduces scratching behavior and EAr edema, as well as decreases levels of IL-1β, IL-4, and IL-6 in EAr skin, in a mouse model of TDI-induced allergic dermatitis at 0.1, 0.25, and 0.5%