Learn More
Cayman Chemical Oclacitinib-13C-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 2750534-84-8, 1 mg
SDP
Catalog No. 502937596
Encompass_Preferred
An internal standard for the quantification of oclacitinib by GC- or LC-MS
Catalog No. 50-293-7596 Supplier Cayman Chemical Supplier No. 288081 MG
An internal standard for the quantification of oclacitinib by GC- or LC-MS