Learn More
Cayman Chemical OclacItInIb-13C-d3 500ug
SDP
Catalog No. 502939017
Encompass_Preferred
An internal standard for the quantification of oclacitinib by GC- or LC-MS
Catalog No. 50-293-9017 Supplier Cayman Chemical Supplier No. 28808500 UG
An internal standard for the quantification of oclacitinib by GC- or LC-MS