Learn More
Cayman Chemical Ocmene 100mg
SDP
Catalog No. 502995296
Encompass_Preferred
A reversible selective allosteric antagonist of mGluR7 that blocks agonist-induced calcium mobilization (IC50 26 nM)
Catalog No. 50-299-5296 Supplier Cayman Chemical Supplier No. 23466100 MG
A reversible selective allosteric antagonist of mGluR7 that blocks agonist-induced calcium mobilization (IC50 26 nM)