Learn More
Cayman Chemical Ocmene 50mg
SDP
Catalog No. 502994691
Encompass_Preferred
A cell-permeable benzoxazolo-sulfonamide compound that blocks human FBPase-1 enzymatic activity by competing at the AMP allosteric binding site (IC50 3 4 UM Ki 1 1 UM) blocks glucose production in starved rat hepatoma cells (IC50 6 6 UM)
Catalog No. 50-299-4691 Supplier Cayman Chemical Supplier No. 2346650 MG
A cell-permeable benzoxazolo-sulfonamide compound that blocks human FBPase-1 enzymatic activity by competing at the AMP allosteric binding site (IC50 3 4 UM Ki 1 1 UM) blocks glucose production in starved rat hepatoma cells (IC50 6 6 UM)