Learn More
Medchemexpress LLC Oct-1-en-3-ol | 3391-86-4 | 100.0% | 128.22 | 50 G
SDP
Catalog No. 5000366448
Encompass_Preferred
Oct-1-en-3-ol is a fatty acid fragrant that acts as a self-stimulating oxylipin messenger. It functions as a signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions. Additionally, Oct-1-en-3-ol has been shown to cause dopamine neuron degeneration by disrupting dopamine handling.
  • Fatty acid fragrant
  • Self-stimulating oxylipin messenger
  • Serves as a signaling molecule in various plant interactions
  • Involved in dopamine neuron degeneration
Catalog No. 50-003-66448 Supplier Medchemexpress LLC Supplier No. HYW01041050G
Oct-1-en-3-ol is a fatty acid fragrant that acts as a self-stimulating oxylipin messenger. It functions as a signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions. Additionally, Oct-1-en-3-ol has been shown to cause dopamine neuron degeneration by disrupting dopamine handling.
  • Fatty acid fragrant
  • Self-stimulating oxylipin messenger
  • Serves as a signaling molecule in various plant interactions
  • Involved in dopamine neuron degeneration