Learn More
Medchemexpress LLC Oct-1-en-3-ol | 3391-86-4 | 99.89% | 128.22 | 1 ML
SDP
Catalog No. 5000367071
Encompass_Preferred
Oct-1-en-3-ol is a fatty acid fragrant and a self-stimulating oxylipin messenger. It acts as a signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions. It can also cause dopamine neuron degeneration by disrupting dopamine handling.
  • Fatty acid fragrant
  • Self-stimulating oxylipin messenger
  • Signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions
  • Causes dopamine neuron degeneration through disruption of dopamine handling
  • Appearance: liquid
  • Color: colorless to light yellow
  • Initial source: plants, microorganisms
Catalog No. 50-003-67071 Supplier Medchemexpress LLC Supplier No. HYW01041010MM/1ML
Oct-1-en-3-ol is a fatty acid fragrant and a self-stimulating oxylipin messenger. It acts as a signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions. It can also cause dopamine neuron degeneration by disrupting dopamine handling.
  • Fatty acid fragrant
  • Self-stimulating oxylipin messenger
  • Signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions
  • Causes dopamine neuron degeneration through disruption of dopamine handling
  • Appearance: liquid
  • Color: colorless to light yellow
  • Initial source: plants, microorganisms