Learn More
Medchemexpress LLC Oct-1-en-3-ol | 3391-86-4 | MFCD00004589 | 99.97% | 128.22 | 100 G
SDP
Catalog No. 5000366472
Encompass_Preferred
Oct-1-en-3-ol, a fatty acid fragrant, is a self-stimulating oxylipin messenger. It serves as a signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions. Oct-1-en-3-ol causes dopamine neuron degeneration through disruption of dopamine handling.
  • Fatty acid fragrant
  • Self-stimulating oxylipin messenger
  • Signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions
  • Causes dopamine neuron degeneration through disruption of dopamine handling
Catalog No. 50-003-66472 Supplier Medchemexpress LLC Supplier No. HYW010410100G
Oct-1-en-3-ol, a fatty acid fragrant, is a self-stimulating oxylipin messenger. It serves as a signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions. Oct-1-en-3-ol causes dopamine neuron degeneration through disruption of dopamine handling.
  • Fatty acid fragrant
  • Self-stimulating oxylipin messenger
  • Signaling molecule in plant cellular responses, plant-herbivore interactions, and plant-plant interactions
  • Causes dopamine neuron degeneration through disruption of dopamine handling