Learn More
Santa Cruz Biotechnology Anti-Oct-3/4 Antibody (C-10), 200 ug/mL
Supplier:  Santa Cruz Biotechnology SC5279
Encompass
Anti-Oct-3/4 Antibody (C-10) is recommended for detection of Oct-3/4 of mouse, rat and human origin by WB, IP, IF, IHC(P), FCM and ELISA; non cross-reactive with Oct-3/4 isoform B.
Catalog No. NC9413705